Hydrocortisone ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A16292.14 A16292.03 A16292.06
A16292.14EA 276 GBP
A16292.14 A16292.03 A16292.06
Hydrocortisone ≥98%
Hydrocortisone

Purity: 98% CAS No.: 50-23-7 Molecular Formula: C21H30O5 Formula Weight: 362.47 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 213-215° Beilstein Reference: 1354819 Merck Reference: 14,4787 MDL No.: MFCD00011654


A natural corticosteroid and anti-inflammatory compound

Formula: C₂₁H₃₀O₅
Boiling Pt: 567 °C (1013 hPa)
Melting Pt: 213…215 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1
Merck Index: 14,4787

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR