Copper(II) acetate monohydrate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A16203.30 A16203.0B A16203.0I
A16203.30EA 29 GBP
A16203.30 A16203.0B A16203.0I
Copper(II) acetate monohydrate ≥98%
Copper (II) acetate monohydrate

Purity: 98+% CAS No.: 6046-93-1 Molecular Formula: C4H6CuO4.H2O Formula Weight: 199.65 (181.64anhy) Physical Form: Powder Density: 1.882 Appearance: Blue-green Odour: Odourless pH: 5.2-5.5 Melting Point: 115° Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00149570

Formula: (H₃CCOO)₂Cu·H₂O
Melting Pt: 115 °C
Density: 1.882
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9,III
Merck Index: 14,2624

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR