Potassium hydroxide ≥85%, flakes

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A16199.36 A16199.0E A16199.0C
A16199.36EA 16.6 GBP
A16199.36 A16199.0E A16199.0C
Potassium hydroxide ≥85%, flakes
Potassium hydroxide

Purity: 85% CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Odour: Odourless pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553

Formula: KOH
Boiling Pt: 1320…1324 °C
Melting Pt: 360 °C
Density: 2.044
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 14,7640

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR