Strontium acetate hemihydrate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Strontium Acetate Hemihydrate

A16149.0E A16149.22 A16149.36
A16149.0EEA 258 GBP
A16149.0E A16149.22 A16149.36
Strontium acetate hemihydrate 98%
Strontium acetate hemihydrate

Purity: 98% CAS No.: 14692-29-6 Molecular Formula: Sr(OOCCH3)2.0.5H2O Formula Weight: 214.72 (205.71anhy) Melting Point: 150° -0.5H2O MDL No.: MFCD00198064

Formula: C₈H₁₄O₉Sr₂
MW: 429.43 g/mol
Melting Pt: 150 °C (-0.5H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00198064
CAS Number: 14692-29-6
EINECS: 208-854-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR