(±)-3-Chloro-1,2-propanediol ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (±)-3-Chloro-1,2-dihydroxy propane

A16143.0E A16143.30 A16143.36
A16143.0EEA 102 GBP
A16143.0E A16143.30 A16143.36
(±)-3-Chloro-1,2-propanediol ≥98%
(±)-3-Chloro-1,2-propanediol

Purity: 98% CAS No.: 96-24-2 Molecular Formula: C3H7ClO2 Formula Weight: 110.54 Flash Point: 137°(278°F) Refractive Index: 1.4800 Melting Point: -40° Boiling Point: 213° Beilstein Reference: 635684 Merck Reference: 14,2145 UN No.: 2689 MDL No.: MFCD00004712

Formula: C₃H₇ClO₂
MW: 110.54 g/mol
Boiling Pt: 213 °C
Melting Pt: –40 °C
Density: 1.322
Flash Pt: 137 °C (278 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004712
CAS Number: 96-24-2
EINECS: 202-492-4
UN: 2689
Merck Index: 13,02164

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR