Lithium perchlorate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A16059.22
A16059.22EA 36.7 GBP
A16059.22
Lithium perchlorate ≥98%
Lithium perchlorate
Formula: ClLiO₄
MW: 106.39 g/mol
Boiling Pt: 430
Melting Pt: 236…237 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011079
CAS Number: 7791-03-9
EINECS: 232-237-2
UN: 1481
ADR: 5.1 II
Merck Index: 13,05561

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR