4-tert-Butylphenol 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A15871.0C A15871.0E A15871.36
A15871.0CEA 249 GBP
A15871.0C A15871.0E A15871.36
4-tert-Butylphenol 99
4-tert-Butylphenol

Purity: 99% CAS No.: 98-54-4 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 113°(235°F) Physical Form: Powder or lumps Density: 0.908 Appearance: White to pale cream Odour: Phenol-like Melting Point: 97-101° Boiling Point: 236-238° Beilstein Reference: 1817334 Merck Reference: 14,1585 UN No.: 3077 MDL No.: MFCD00002367

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 236…238 °C
Melting Pt: 97…101 °C
Density: 0.908
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 3077
ADR: 9 III
Merck Index: 13,01584

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR