Phenol 99+%, detached crystals

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

A15760.0E A15760.22 A15760.36
A15760.0EEA 96.6 GBP
A15760.0E A15760.22 A15760.36
Phenol 99+%, detached crystals
Phenol

Purity: 99+% CAS No.: 108-95-2 Molecular Formula: C6H6O Formula Weight: 94.11 Flash Point: 79°(175°F) Odour: Sweet Boiling Point: 181° Beilstein Reference: 969616 Merck Reference: 14,7241 UN No.: 1671 MDL No.: MFCD00002143

Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181 °C
Melting Pt: 40…43 °C
Density: 1.071
Flash Pt: 79 °C (175 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
Merck Index: 14,7241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR