2-Bromobenzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (2-Bromophenyl)methanol

A15700.09 A15700.18
A15700.09EA 30.8 GBP
A15700.09 A15700.18
2-Bromobenzyl alcohol ≥98%
2-Bromobenzyl alcohol

Purity: 98% CAS No.: 18982-54-2 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 78-82° Beilstein Reference: 2243418

Formula: C₇H₇BrO
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004600
CAS Number: 18982-54-2
EINECS: 242-719-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR