Itaconic acid ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A15566.0B A15566.0I A15566.30
A15566.0BEA 31.9 GBP
A15566.0B A15566.0I A15566.30
Itaconic acid ≥99%
Itaconic acid

Purity: 99% CAS No.: 97-65-4 Molecular Formula: C5H6O4 Formula Weight: 130.10 Melting Point: 165-169° Boiling Point: 268° Merck Reference: 14,5242 MDL No.: MFCD00004260

Formula: C₅H₆O₄
Boiling Pt: 268 °C
Melting Pt: 165…169 °C
Density: 1.630
Storage Temperature: Ambient
MDL Number: MFCD00004260
CAS Number: 97-65-4
EINECS: 202-599-6
Merck Index: 14,5242

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR