Saccharin sodium hydrate ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: o-Benzoic sulphimide sodium salt hydrate, Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide hydrate, Saccharin, sodium salt hydrate

A15530.0B A15530.0I A15530.30
A15530.0BEA 90.4 GBP
A15530.0B A15530.0I A15530.30
Saccharin sodium hydrate ≥99%
Saccharin sodium hydrate

Purity: 99% CAS No.: 82385-42-0 Molecular Formula: C7H4NNaO3S.xH2O Formula Weight: 205.16(anhy) Melting Point: 300° Beilstein Reference: 3599229 Merck Reference: 14,8312 MDL No.: MFCD00151213

Formula: C₇H₅NNaO₃S·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149605
CAS Number: 82385-42-0
EINECS: 204-886-1
Merck Index: 14,8312

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR