Ammonium dihydrogen phosphate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

A15283.0C A15283.0E A15283.36 A15283.0G
A15283.0CEA 316 GBP
A15283.0C A15283.0E A15283.36 A15283.0G
Ammonium dihydrogen phosphate ≥98%
Ammonium dihydrogen phosphate

Purity: 98% CAS No.: 7722-76-1 Molecular Formula: NH4H2PO4 Formula Weight: 115.03 Odour: Odourless Melting Point: 198-204° MDL No.: MFCD00003396

Formula: NH₄H₂PO₄
Melting Pt: 198…204 °C
Density: 1.803
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 14,543

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR