4-Butylbenzoyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A15082.06 A15082.14
A15082.06EA 37.7 GBP
A15082.06 A15082.14
4-Butylbenzoyl chloride ≥98%
4-Butylbenzoyl chloride

Purity: 98% CAS No.: 28788-62-7 Molecular Formula: C11H13ClO Formula Weight: 196.68 Flash Point: 110°(230°F) Refractive Index: 1.5351 Boiling Point: 145-147°/11mm UN No.: 3265 MDL No.: MFCD00000699

Formula: C₁₁H₁₃ClO
Boiling Pt: 145…147 °C (11 mmHg)
Density: 1.051
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000699
CAS Number: 28788-62-7
EINECS: 249-224-2
UN: 3265
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR