Benzyltrimethylammonium hydroxide 40% (w/w) in methanol

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

A14927.0F A14927.AE A14927.AP
A14927.0FEA 188 GBP
A14927.0F A14927.AE A14927.AP
Benzyltrimethylammonium hydroxide 40% (w/w) in methanol
Benzyltrimethylammonium hydroxide

CAS No.: 100-85-6 Molecular Formula: C10H17NO Formula Weight: 167.25 Flash Point: 15°(59°F) Refractive Index: 1.4270 Odour: Ammonia-like Boiling Point: 65° Beilstein Reference: 3917256 UN No.: 2924 MDL No.: MFCD00008281

Formula: C₁₀H₁₇NO
Boiling Pt: 65 °C
Density: 0.920
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008281
CAS Number: 100-85-6
EINECS: 202-895-5
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR