Isobutyl acetate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Methylpropyl acetate

A14892.0F A14892.AK A14892.AP
A14892.0FEA 52.3 GBP
A14892.0F A14892.AK A14892.AP
Isobutyl acetate ≥98%
Isobutyl acetate

Purity: 98% CAS No.: 110-19-0 Molecular Formula: CH3CO2CH2CH(CH3)2 Formula Weight: 116.16 Flash Point: 18°(64°F) Physical Form: Liquid Density: 0.870 Refractive Index: 1.3895 Odour: Fruit-like Melting Point: -99° Boiling Point: 116-117° Beilstein Reference: 1741909 Merck Reference: 14,5130 UN No.: 1213 MDL No.: MFCD00008932

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 116…117 °C
Melting Pt: –99 °C
Density: 0.870
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008932
CAS Number: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3 II
Merck Index: 13,05147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR