n-Dodecane ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Dodecane

A14834.0F A14834.AE A14834.AP A14834.PI
A14834.0FEA 333 GBP
A14834.0F A14834.AE A14834.AP A14834.PI
n-Dodecane ≥99%
n-Dodecane

Purity: 99+% CAS No.: 112-40-3 Molecular Formula: CH3(CH2)10CH3 Formula Weight: 170.34 Flash Point: 71°(159°F) Physical Form: Liquid Density: 0.749 Refractive Index: 1.4220 Odour: Petrol-like Melting Point: -10° Boiling Point: 215-217° Beilstein Reference: 1697175 MDL No.: MFCD00008969

Formula: H₃C(CH₂)₁₀CH₃
MW: 170.34 g/mol
Boiling Pt: 215…217 °C
Melting Pt: –10 °C
Density: 0.749
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008969
CAS Number: 112-40-3
EINECS: 203-967-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d3d75a63f52fbe2a8be1681d9233be6dc0928&vid=3268fe23453ad703df2782bd3c7ba6b939ba00cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR