Cytosine ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A14731.14 A14731.06 A14731.22
A14731.14EA 122 GBP
A14731.14 A14731.06 A14731.22
Cytosine ≥98%
Cytosine

Purity: 98+% CAS No.: 71-30-7 Molecular Formula: C4H5N3O Formula Weight: 111.10 Physical Form: Powder Appearance: White Melting Point: 300° Beilstein Reference: 2637 Merck Reference: 14,2795 MDL No.: MFCD00006034

Formula: C₄H₅N₃O
Melting Pt: >300 °C
Density: 0.99771 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5
Merck Index: 14,2795

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR