Sodium-1-decanesulphonate, (max. 1,5% H₂O) 99% (dry weight)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

A14638.06 A14638.14 A14638.22
A14638.06EA 40.3 GBP
A14638.06 A14638.14 A14638.22
Sodium-1-decanesulphonate, (max. 1,5% H₂O) 99% (dry weight)
Sodium 1-decanesulphonate

Purity: 99% CAS No.: 13419-61-9 Molecular Formula: CH3(CH2)9SO3Na Formula Weight: 244.33 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 300° Beilstein Reference: 3918920 MDL No.: MFCD00007526


Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.

Formula: C₁₀H₂₁NaO₃S
MW: 244.33 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR