Hydrobromic acid solution_SC 33% w/w 45% w/v acetic acid

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydrogen bromide

A14475.0F A14475.AE A14475.AP
A14475.0FEA 374 GBP
A14475.0F A14475.AE A14475.AP
Hydrobromic acid solution_SC 33% w/w 45% w/v acetic acid
Hydrogen bromide

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds


It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

Formula: HBr
MW: 80.91 g/mol
Boiling Pt: -66.7
Melting Pt: -86
Density: 1.40
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 37348-16-6
EINECS: 233-113-0
UN: 3265
ADR: 8 II
Merck Index: 14,4778

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR