(±)-Linalool 97

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol

A14424.0F A14424.AE A14424.AP
A14424.0FEA 287 GBP
A14424.0F A14424.AE A14424.AP
(±)-Linalool 97
(±)-Linalool

Purity: 97% CAS No.: 78-70-6 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 76°(168°F) Physical Form: Liquid Density: 0.865 Refractive Index: 1.4620 Appearance: Colourless Boiling Point: 199-200° Beilstein Reference: 1721488 Merck Reference: 14,5495 MDL No.: MFCD00008906

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 199…200 °C
Density: 0.865
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008906
CAS Number: 78-70-6
EINECS: 201-134-4
Merck Index: 13,05517

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR