Salicyl alcohol ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A14410.0B A14410.18 A14410.30
A14410.0BEA 560 GBP
A14410.0B A14410.18 A14410.30
Salicyl alcohol ≥99%
Salicyl alcohol

Purity: 99% CAS No.: 90-01-7 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 83-87° Beilstein Reference: 1907195 Merck Reference: 14,8325 MDL No.: MFCD00004617

Formula: C₇H₈O₂
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 83…87 °C
Density: 1.1613 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004617
CAS Number: 90-01-7
EINECS: 201-960-5
Merck Index: 14,8325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR