Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A14315.22 A14315.36 A14315.0E
A14315.22EA 24.4 GBP
A14315.22 A14315.36 A14315.0E
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Polyvinylpyrrolidone

CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Merck Reference: 14,7697 MDL No.: MFCD00149016

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR