3,4,5-Trimethoxybenzoic acid ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A14274.22
A14274.22EA 36.6 GBP
A14274.22
3,4,5-Trimethoxybenzoic acid ≥99%
3,4,5-Trimethoxybenzoic acid

Purity: 99+% CAS No.: 118-41-2 Molecular Formula: C10H12O5 Formula Weight: 212.20 Melting Point: 168-171° Boiling Point: 225-226°/10mm MDL No.: MFCD00002501

Formula: C₁₀H₁₂O₅
Boiling Pt: 225…226 °C (10 mmHg)
Melting Pt: 168…171 °C
Density: 1.31 g/cm³ (24.95 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002501
CAS Number: 118-41-2
EINECS: 204-248-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR