3-Acetylpyridine ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A14246.14 A14246.22 A14246.36
A14246.14EA 25.6 GBP
A14246.14 A14246.22 A14246.36
3-Acetylpyridine ≥98%
3-Acetylpyridine

Purity: 98% CAS No.: 350-03-8 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 104°(219°F) Refractive Index: 1.5350 Odour: Characteristic Melting Point: 12-13° Boiling Point: 218-220° Beilstein Reference: 107751 Merck Reference: 14,6116 UN No.: 2810 MDL No.: MFCD00006396

Formula: C₇H₇NO
Boiling Pt: 218…220 °C
Melting Pt: 12…13 °C
Density: 1.106
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,II
Merck Index: 14,6116

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR