2-Mercaptobenzothiazole ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

A14086.09 A14086.0E A14086.22 A14086.36
A14086.09EA 16 GBP
A14086.09 A14086.0E A14086.22 A14086.36
2-Mercaptobenzothiazole ≥97%
2-Mercaptobenzothiazole

Purity: 97% CAS No.: 149-30-4 Molecular Formula: C7H5NS2 Formula Weight: 167.25 Flash Point: 243°(469°F) Melting Point: 178-182° Beilstein Reference: 119484 Merck Reference: 14,5868 UN No.: 3077 MDL No.: MFCD00005781

Formula: C₇H₅NS₂
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 178…182 °C
Density: 1.420
Flash Pt: 243 °C (469 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 14,5868

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR