Ethyl hexanoate ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A13985.0E A13985.22 A13985.36
A13985.0EEA 165 GBP
A13985.0E A13985.22 A13985.36
Ethyl hexanoate ≥99%
Ethyl hexanoate

Purity: 99% CAS No.: 123-66-0 Molecular Formula: CH3(CH2)4CO2CH2CH3 Formula Weight: 144.21 Flash Point: 49°(121°F) Refractive Index: 1.4075 Melting Point: -67° Boiling Point: 166° Merck Reference: 14,3777 UN No.: 3272 MDL No.: MFCD00009511

Formula: C₈H₁₆O₂
Boiling Pt: 166 °C
Melting Pt: –67 °C
Density: 0.873
Flash Pt: 49 °C (121 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009511
CAS Number: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 14,3777

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR