4-Fluoro-o-phenylenediamine 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine

A13977.06 A13977.14
A13977.06EA 46.8 GBP
A13977.06 A13977.14
4-Fluoro-o-phenylenediamine 97%
4-Fluoro-o-phenylenediamine

Purity: 97% CAS No.: 367-31-7 Molecular Formula: C6H7FN2 Formula Weight: 126.13 Melting Point: 91-95° Beilstein Reference: 2081075 UN No.: 2811 MDL No.: MFCD00042228

Formula: C₆H₇FN₂
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00042228
CAS Number: 367-31-7
EINECS: 206-691-7
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR