Potassium-tert-butoxide ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

A13947.30 A13947.0B A13947.14 A13947.22 A13947.36 AB119358.0100
A13947.30EA 65.2 GBP
A13947.30 A13947.0B A13947.14 A13947.22 A13947.36 ABCRAB119358.0100
Potassium-tert-butoxide ≥97%
Potassium-tert-butoxide

Purity: 97% CAS No.: 865-47-4 Molecular Formula: C4H9KO Formula Weight: 112.22 Melting Point: ca 255° dec. Beilstein Reference: 3556712 UN No.: 3206 MDL No.: MFCD00012162

Formula: C₄H₉KO
Melting Pt: ∼ 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR