β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A1393.0001 A1393.0010
A1393.0001EA 102 GBP
A1393.0001 A1393.0010
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number: MFCD00036200
CAS Number: 606-68-8
EINECS: 210-123-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR