Benzylphosphonic acid ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A13850.03 A13850.06 A13850.14
A13850.03EA 26.6 GBP
A13850.03 A13850.06 A13850.14
Benzylphosphonic acid ≥97%
Benzylphosphonic acid

Purity: 97% CAS No.: 6881-57-8 Molecular Formula: C6H5CH2PO3H2 Formula Weight: 172.11 Melting Point: 166-172° Beilstein Reference: 2519318 UN No.: 3261 MDL No.: MFCD00039519

Formula: C₇H₉O₃P
MW: 172.11 g/mol
Melting Pt: 166…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00039519
CAS Number: 6881-57-8
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR