Quinaldine 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Methylquinoline

A13412.14 A13412.22 A13412.36
A13412.14EA 22.5 GBP
A13412.14 A13412.22 A13412.36
Quinaldine 96%
Quinaldine

Used in preparation of oil-soluble dyes


It is an anaesthetic used in fish transportation. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.


Caution: Air sensitive. Light sensitive. Store under argon.

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 247…248 °C
Melting Pt: –2 °C
Density: 1.058
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006756
CAS Number: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Order Now

Specification Test Results

Appearance (Colour) Clear colorless to yellow to brown
Form Liquid
Assay (GC) ≥95,0%
Refractive Index 1,6080 - 1,6145 @ 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR