3-Aminopropionitrile 98% Stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: β-Aminopropionitrile, 3-Aminopropanenitrile

Stabilisation: K2CO3

A13043.03 A13043.06 A13043.14
A13043.03EA 22.4 GBP
A13043.03 A13043.06 A13043.14
3-Aminopropionitrile 98% Stabilised
3-Aminopropionitrile

Stabilized with potassium carbonate


Purity: 98% CAS No.: 151-18-8 Molecular Formula: C3H6N2 Formula Weight: 70.10 Storage Temperature: 2°C to 8°C Refractive Index: 1.4375 Odour: Amine-like Boiling Point: 79-81°/16mm Beilstein Reference: 1698848 Merck Reference: 14,469 UN No.: 2735 MDL No.: MFCD00014820

Formula: C₃H₆N₂
MW: 70.10 g/mol
Boiling Pt: 79…81 °C (16 mmHg)
Density: 0.952
Storage Temperature: Refrigerator
MDL Number: MFCD00014820
CAS Number: 151-18-8
EINECS: 205-786-0
UN: 2735
ADR: 8 III
Merck Index: 14,469

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR