3,5-Dichloro-4-hydroxybenzoic acid ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A12923.14 A12923.06 A12923.22
A12923.14EA 47.9 GBP
A12923.14 A12923.06 A12923.22
3,5-Dichloro-4-hydroxybenzoic acid ≥97%
3,5-Dichloro-4-hydroxybenzoic acid

Purity: 97% CAS No.: 3336-41-2 Molecular Formula: C7H4Cl2O3 Formula Weight: 207.01 Melting Point: 265-270° Beilstein Reference: 2616297

Formula: C₇H₄Cl₂O₃
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 265…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00002550
CAS Number: 3336-41-2
EINECS: 222-071-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR