4-Chloroaniline ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A12879.30 A12879.0B A12879.0I
A12879.30EA 35.2 GBP
A12879.30 A12879.0B A12879.0I
4-Chloroaniline ≥98%
4-Chloroaniline

Purity: 98% CAS No.: 106-47-8 Molecular Formula: C6H6ClN Formula Weight: 127.57 Flash Point: 120°(248°F) Melting Point: 67-72° Boiling Point: 232° Beilstein Reference: 471359 Merck Reference: 14,2118 UN No.: 2018 MDL No.: MFCD00007835

Formula: C₆H₆ClN
Boiling Pt: 232 °C
Melting Pt: 67…72 °C
Density: 1.430
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 14,2118

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR