α,α,α-Trifluoro-m-cresol ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride

A12784.06 A12784.14 A12784.22
A12784.06EA 15.5 GBP
A12784.06 A12784.14 A12784.22
α,α,α-Trifluoro-m-cresol ≥98%
α,α,α-Trifluoro-m-cresol

Purity: 98+% CAS No.: 98-17-9 Molecular Formula: C7H5F3O Formula Weight: 162.11 Flash Point: 74°(165°F) Refractive Index: 1.4580 Odour: Phenol-like Melting Point: -2° Boiling Point: 177-178° Beilstein Reference: 2045663 UN No.: 3265 MDL No.: MFCD00002299

Formula: C₇H₅F₃O
Boiling Pt: 177…178 °C
Melting Pt: –2 °C
Density: 1.336
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR