4,4'-Dichlorobenzophenone ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4,4-Dichlorobenzophenone, 4,4-DBP, Bis(4-chlorophenyl) ketone

A12608.14 A12608.22 A12608.36
A12608.14EA 13 GBP
A12608.14 A12608.22 A12608.36
4,4'-Dichlorobenzophenone ≥99%
4,4'-Dichlorobenzophenone

Purity: 99% CAS No.: 90-98-2 Molecular Formula: C13H8Cl2O Formula Weight: 251.11 Melting Point: 144-146° Boiling Point: 352-354° MDL No.: MFCD00000623

Formula: C₁₃H₈Cl₂O
Boiling Pt: 352…354 °C
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00000623
CAS Number: 90-98-2
EINECS: 202-030-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR