Ethyl-3-bromopropionate ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A12402.14 A12402.22 A12402.36 AB148814.0025
A12402.14EA 35.3 GBP
A12402.14 A12402.22 A12402.36 ABCRAB148814.0025
Ethyl-3-bromopropionate ≥99%
Ethyl 3-bromopropionate

Purity: 99% CAS No.: 539-74-2 Molecular Formula: BrCH2CH2CO2CH2CH3 Formula Weight: 181.03 Flash Point: 79°(174°F) Refractive Index: 1.4520 Boiling Point: 67-71°/10mm Beilstein Reference: 1700933 MDL No.: MFCD00000251

Formula: C₅H₉BrO₂
Boiling Pt: 67…71 °C (10 mmHg)
Density: 1.416
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000251
CAS Number: 539-74-2
EINECS: 208-724-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR