Zirconyl chloride octahydrate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Zirconium(IV) oxide chloride octahydrate, Zirconium dichloride oxide octahydrate, Zirconium(IV) oxychloride octahydrate

A12342.0E A12342.22 A12342.36
A12342.0EEA 744 GBP
A12342.0E A12342.22 A12342.36
Zirconyl chloride octahydrate ≥98%
Zirconyl chloride octahydrate

Purity: 98% CAS No.: 13520-92-8 Molecular Formula: ZrOCl2.8H2O Formula Weight: 322.25 (178.13anhy) Odour: Acidic Melting Point: 150° dec. UN No.: 3260 MDL No.: MFCD00149898

Formula: Cl₂OZr·8H₂O
Melting Pt: 150 °C (dec.)
Density: 1.91
Storage Temperature: Ambient
MDL Number: MFCD00149898
CAS Number: 13520-92-8
EINECS: 231-717-9
UN: 3260
ADR: 8,II
Merck Index: 14,10184

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR