Sodium nitrate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A12327.0C A12327.0E A12327.36
A12327.0CEA 190 GBP
A12327.0C A12327.0E A12327.36
Sodium nitrate ≥98%
Sodium nitrate

Purity: 98+% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Odour: Odourless Melting Point: ca 308° UN No.: 1498 MDL No.: MFCD00011119

Formula: NaNO₃
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: ∼308 °C
Density: 2.26
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 14,8647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR