1,1,3,3-Tetramethylguanidine ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N,N,N',N'-Tetramethylguanidine

A12314.AC A12314.AE A12314.AP
A12314.ACEA 20 GBP
A12314.AC A12314.AE A12314.AP
1,1,3,3-Tetramethylguanidine ≥99%
1,1,3,3-Tetramethylguanidine

Purity: 99% CAS No.: 80-70-6 Molecular Formula: C5H13N3 Formula Weight: 115.18 Flash Point: 60°(140°F) Refractive Index: 1.4690 Melting Point: -30° Boiling Point: 162° Beilstein Reference: 969608 UN No.: 2920 MDL No.: MFCD00008323

Formula: C₅H₁₃N₃
Boiling Pt: 162 °C
Melting Pt: < –30 °C
Density: 0.918
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR