Methylammonium chloride 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: MACl, Methylammonium chloride, Aminomethane hydrochloride, Methylamine hydrochloride

A12291.22 A12291.36 A12291.PG
A12291.22EA 19.4 GBP
A12291.22 A12291.36 A12291.PG
Methylammonium chloride 99%
Methylammonium chloride

The HCl salt is in the form of white deliquescent crystals, and is frequently substitutable for the aq. soln. in many synthesis preparations. Methylamines are used directly as catalysts or as raw materials to produce other compounds with catalytic activity. Fuel additives are used to improve engine performance in a variety of ways. Trimethylamine is used to make paper chemicals. The manufacture of intermediates to make pharmaceuticals is one of the most diverse uses of methylamines


Caution: Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.

Formula: CH₆ClN
Boiling Pt: 225…230 °C (15 mmHg)
Melting Pt: 230…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00012849
CAS Number: 593-51-1
EINECS: 209-795-0
Merck Index: 14,6014

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR