Sudan IV

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol

A12181.22 A12181.14
A12181.22EA 39.9 GBP
A12181.22 A12181.14
Sudan IV
Sudan IV

CAS No.: 85-83-6 Molecular Formula: C24H20N4O Formula Weight: 380.45 Melting Point: ca 185° dec. Merck Reference: 14,8393 MDL No.: MFCD00003893

Formula: C₂₄H₂₀N₄O
MW: 380.45 g/mol
Melting Pt: ∼ 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003893
CAS Number: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR