4,4'-Bis(dimethylamino)benzophenone ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A11889.22 A11889.36
A11889.22EA 31.7 GBP
A11889.22 A11889.36
4,4'-Bis(dimethylamino)benzophenone ≥98%
4,4'-Bis(dimethylamino)benzophenone

Purity: 98% CAS No.: 90-94-8 Molecular Formula: C17H20N2O Formula Weight: 268.36 Flash Point: 220°(428°F) Melting Point: 174-176° Boiling Point: 360° Beilstein Reference: 790733 Merck Reference: 14,6177 MDL No.: MFCD00008312

Formula: C₁₇H₂₀N₂O
Boiling Pt: >360 °C
Melting Pt: 174…176 °C
Flash Pt: 220 °C (428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008312
CAS Number: 90-94-8
EINECS: 202-027-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,6177

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR