Sodium thiocyanate 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Sodium rhodanide

A11867.0B A11867.0I A11867.30
A11867.0BEA 67.5 GBP
A11867.0B A11867.0I A11867.30
Sodium thiocyanate 96%
Sodium thiocyanate

Purity: 96% CAS No.: 540-72-7 Molecular Formula: NaSCN Formula Weight: 81.07 Odour: Odourless pH: 6-8 Melting Point: 287° Beilstein Reference: 3594965 Merck Reference: 14,9327 MDL No.: MFCD00011123

Formula: NaSCN
MW: 81.07 g/mol
Melting Pt: 287
Density: 1.74
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 208-754-4
Merck Index: 13,09401

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR