N-Methylformanilide ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A11829.0C A11829.0E A11829.30 A11829.36
A11829.0CEA 893 GBP
A11829.0C A11829.0E A11829.30 A11829.36
N-Methylformanilide ≥99%
N-Methylformanilide

Purity: 99% CAS No.: 93-61-8 Molecular Formula: C8H9NO Formula Weight: 135.17 Flash Point: 126°(258°F) Refractive Index: 1.5610 Melting Point: 9-13° Boiling Point: 242-244° Beilstein Reference: 636496 MDL No.: MFCD00003283

Formula: C₈H₉NO
Boiling Pt: 242…244 °C
Melting Pt: 9…13 °C
Density: 1.095
Flash Pt: 126 °C (258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003283
CAS Number: 93-61-8
EINECS: 202-262-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR