Sodium octyl sulphate ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Octyl sulphate, sodium salt, Octyl sulphuric acid salt

A11786.03 A11786.06 A11786.14
A11786.03EA 33.7 GBP
A11786.03 A11786.06 A11786.14
Sodium octyl sulphate ≥99%
Sodium octyl sulphate

Purity: 99% CAS No.: 142-31-4 Molecular Formula: CH3(CH2)7OSO3Na Formula Weight: 232.28 Melting Point: ca 195° dec. Beilstein Reference: 3658635

Formula: C₈H₁₇NaO₄S
Melting Pt: ∼195 °C (dec.)
Density: 1.266 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007470
CAS Number: 142-31-4
EINECS: 205-535-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR