Ethanolamine ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A11697.0E A11697.36
A11697.0EEA 54 GBP
A11697.0E A11697.36
Ethanolamine ≥98%
Ethanolamine

Purity: 98+% CAS No.: 141-43-5 Molecular Formula: C2H7NO Formula Weight: 61.08 Flash Point: 93°(199°F) Refractive Index: 1.4540 Odour: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183

Formula: NH₂CH₂CH₂OH
Boiling Pt: 169…170 °C
Melting Pt: 10 °C
Density: 1.012
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
Merck Index: 14,3727
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR