Semicarbazide hydrochloride 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

A11668.0E A11668.22 A11668.36
A11668.0EEA 308 GBP
A11668.0E A11668.22 A11668.36
Semicarbazide hydrochloride 99
Semicarbazide hydrochloride

Purity: 99% CAS No.: 563-41-7 Molecular Formula: CH4N3O.HCl Formula Weight: 111.53 Physical Form: Crystalline Appearance: White Odour: Odourless pH: 1.5 Melting Point: ca 176° dec. Beilstein Reference: 3593642 Merck Reference: 14,8444 UN No.: 2811 MDL No.: MFCD00013009


Urease substrate and MAO inhibitor

Formula: CH₆ClN₃O
MW: 111.53 g/mol
Melting Pt: ∼ 176 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013009
CAS Number: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1 III
Merck Index: 12,08588

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR