N-Boc-glycine (boc-gly-OH) ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A11579.14 A11579.22
A11579.14EA 45.1 GBP
A11579.14 A11579.22
N-Boc-glycine (boc-gly-OH) ≥98%
N-Boc-glycine (Boc-Gly-OH)

Purity: 98+% CAS No.: 4530-20-5 Molecular Formula: (CH3)3COCONHCH2CO2H Formula Weight: 175.18 Melting Point: 87-90° Beilstein Reference: 1101514

Formula: C₇H₁₃NO₄
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00002690
CAS Number: 4530-20-5
EINECS: 224-864-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR