trans-2-Hexenal 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A11513.14 A11513.22
A11513.14EA 27.5 GBP
A11513.14 A11513.22
trans-2-Hexenal 96%
trans-2-Hexenal

Purity: 96% CAS No.: 6728-26-3 Molecular Formula: C6H10O Formula Weight: 98.15 Storage Temperature: 2°C to 8°C Flash Point: 38°(100°F) Refractive Index: 1.4470 Odour: Sweet Boiling Point: 37-38°/8mm Beilstein Reference: 1699684 UN No.: 1988 MDL No.: MFCD00007008

Formula: C₆H₁₀O
Boiling Pt: 37…38 °C (8 mmHg)
Density: 0.848
Flash Pt: 38 °C (100 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007008
CAS Number: 6728-26-3
EINECS: 229-778-1
UN: 1988
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR